Caplugs evergreen cassette storage drawers are sturdy plastic storage library units that hold approximately 250 embedding rings, 400 processing/embedding cassettes, or combinations of both.
- Constructed of High Impact Polystyrene (HIP) for strength and flatness
- Inner drawer is removable and comes with large ID labels
- Units are stackable
Different color file cabinets allow you to customize your own storage system or color code different types of tissue and/or biopsy cassettes.