Order Entry
United States
ContactUsLinkComponent
 
PrimeStore® Molecular Transport Medium (MTM), SiO₂ Medical Products
Catalog # 76545-528
Supplier:  Avantor
CAS Number:  
undefined
PrimeStore® Molecular Transport Medium (MTM), SiO₂ Medical Products
Catalog # 76545-528
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    PP tube
  • Environmentally Preferable:
  • Cat. No.:
    76545-528
  • Fill volume:
    2 ml
  • Tube size:
    0.648' Ext.Ø×3.850' L
  • Volume:
    12 ml

Specifications

About this item

The product is a ready-to-use capped and labeled 12 ml tube that is filled with 2 ml of Molecular Transport Medium (MTM) solution.

  • FDA registered Class II Medical Device
  • Threaded with red pre-assembled screw caps
  • Leak proof
  • 2-year shelf-life from date of manufacture

PrimeStore MTM is intended for the stabilization, transportation and inactivation of infectious unprocessed nasal washes suspected of containing Influenza A virus RNA. PrimeStore MTM is also intended for the stablization, transportation and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium Tuberculosis DNA from human samples.