United Scientific™ Variable Volume Fully Autoclavable Micropipettes
Supplier: United Scientific Supplies
Certificates
About this item
Variable volume micropipetttes with tip ejectors.
- Fully autoclavable micropipette with volume range
- Designed with click-stop volume control, built-in tip ejectors, autoclavable cone tips and comfortable grips
The variable volume micropipettes feature built-in tip ejectors and autoclavable tip cones. They are offered in nine volume ranges, as detailed below. They work on a click stop digital system, are easy to calibrate and maintain, and easy to disassemble for autoclaving.
Certifications: Manufactured in an ISO 9001:2008 certified facility, these pipettes are highly accurate. Each pipette is individually calibrated according to ISO 8655 standards. A calibration certificate is provided with each pipette.
Frequently Bought Together