ContactUsLinkComponent
Innovating Science® Presumptive Blood Test Kit
Innovating Science® Presumptive Blood Test Kit
Catalog # 470343-394
Supplier:  Ward's Science
undefined
Innovating Science® Presumptive Blood Test Kit
Catalog # 470343-394
Supplier:  Ward's Science
Supplier Number:  IS9002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Presumptive blood test kit
  • Environmentally Preferable:
  • Cat. No.:
    470343-394
  • No. of uses:
    30
Specifications
About this item
Learn about how to test for the presence of blood on materials.
  • Students will test materials for the presence of blood.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-3
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Test for the presence of blood on materials using phenolphthalein. The test will not distinguish between animal and human blood. Further serology tests are needed. Kit contains instructions, blood standard and reagents to complete 30 tests.
Delivery: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Phenolphthalein and Methanol, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.