3-Prong Clamp with Flexible Extension Rod
Catalog # 470339-746
Supplier: United Scientific Supplies
Specifications
- Description:3-prong clamp with flexible extension rod
- Environmentally Preferable:
- Cat. No.:470339-746
Specifications
About this item
Sturdy flexible support arm with clamp.
- Flexible arm
- Holds up to 2 pounds
This flexible arm is extremely versatile and can be placed in virtually any position or angle. Includes a 3-prong clamp head, and nickel-plated flex arm 305 mm (12") in length. The Lab-Frame Connector securely attaches to support stands, lab-frames, or any support rod up to 19 mm (0.75") diameter.