Specifications
- Capacity:6x 1300 ml
- Description:Alcohol-free hand sanitizer, refill
- Environmentally Preferable:
- Cat. No.:76547-798
Specifications
About this item
Soft Care® Instant Hand Sanitizer AF is a benzalkonium chloride based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protected against irritation.
- Alcohol-free formulation for use where alcohol-based hand sanitizers are a concern
- Safe, quick and effective hand rub helps reduce the spread of germs
- Skin softening emollients leave hands soft and with no sticky residues
- Dye-free, reducing potential skin irritation and allergic reactions for users with sensitive skin
- NSF E-3 certified for use in food handling establishments
Dye-free and alcohol-free.
It is a preferred option for those facilities who do not desire alcohol based products.