Hayward 3-Way Electrically Actuated Ball Valves
NEMA 4X polypropylene housing for harsh operating conditions.
- Thermal overload circuit protects the motor from burnout and overheating
These Hayward 3-way electrically actuated ball valves are ideal for use in harsh applications. All valves feature Viton® o-rings. Choose between valves with PVC or CPVC body construction.
- Environmentally Preferable:
Frequently Bought Together