Burkert® Solenoid-Operated VDC Valves
Supplier: VWR International
Certificates
About this item
Fast-acting long life solenoid valves.
- Constructed of durable stainless steel (SS) and fluoropolymer elastomer (FPM)
Choose from two- or three-way valves, with the port, orifice, and voltage your applciation requires.
- Environmentally Preferable:
Frequently Bought Together