Order Entry
United States
ContactUsLinkComponent
Pregnant Mare Serum Gonadotropin, 1000 IU
Pregnant Mare Serum Gonadotropin, 1000 IU
Catalog # 103737-140
Supplier:  BioVendor
CAS Number:  
undefined
Pregnant Mare Serum Gonadotropin, 1000 IU
Catalog # 103737-140
Supplier:  BioVendor
Supplier Number:  RP1782721000
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Native
  • Source:
    Pregnant mare serum
  • Species:
    Horse
  • Size:
    1000 IU
  • Environmentally Preferable:
  • Storage Conditions:
    Lyophilized PMSG although stable at room temperature for 3 weeks, should be stored between 2…8 °C
  • Reconstitution Instructions:
    It is recommended to reconstitute the lyophilized PMSG in sterile 18 MΩ-cm H₂O at a concentration of 1000 IU/ml, which can then be further diluted to other aqueous solutions
  • Protein Synonyms:
    PMSG
  • Protein/Peptide Name:
    Pregnant Mare Serum Gonadotropin (PMSG)
  • Molecular Weight:
    43 - 63 kDa
  • Formulation:
    The PMSG was lyophilized with no additives
  • Shipping Temperature:
    At ambient temperature
  • Cat. No.:
    103737-140

Specifications

About this item

PMSG is a complex glycoprotein obtained from the serum of pregnant mares.

  • Serum of pregnant mares
  • Sterile Filtered White lyophilized (freeze-dried) powder
  • The PMSG was lyophilized with no additives
  • It is recommended to reconstitute the lyophilized PMSG in sterile 18 MΩ-cm H₂O at aconcentration of 1000 IU/ml, which can then be further diluted to other aqueous solutions
  • Lyophilized PMSG although stable at room temperature for three weeks, should be stored between 2 to 8 °C

This 43–63 kda protein is capable of supplementing and being substituted for the follicle stimulating gonadotrophin and interstitial cell-stimulating hormone of the anterior pituitary gland in both the male and female. Thus PMSG-Intervet stimulates development of the ovarian follicle in the female.

Caution: Research use only.