ContactUsLinkComponent
SCape® CO/CBRN 30 Escape Respirator, ILC Dover
SCape® CO/CBRN 30 Escape Respirator, ILC Dover
Catalog # 76533-512
Supplier:  ILC DOVER LP
undefined
SCape® CO/CBRN 30 Escape Respirator, ILC Dover
Catalog # 76533-512
Supplier:  ILC DOVER LP
Supplier Number:  6000-11300-03
Specifications
  • Description:
    SCape® CO/CBRN 30 escape respirator
  • Size:
    Universal
  • Environmentally Preferable:
  • Cat. No.:
    76533-512
Specifications
About this item
The SCape® CO/CBRN30 from ILC Dover is the most advanced, FM and NIOSH-approved air-purifying escape respirator on the market. It provides mission critical protection from carbon monoxide, chemical, biological, radiological, and nuclear contaminants.
  • Delivers reliable protection with minimal training
  • Eases physiological burden on user
  • Reduces claustrophobic feelings associated with other hoods
  • Aids breathing for those with respiratory issues (e.g. Asthma)
A 'one size fits all' design eliminates the need for fit testing, as well as the need to store multiple sizes. The clear hood and large visor allows users to perform a wide range of tasks, including reading and driving. Additionally, this permits total facial recognition. The elimination of headbands and nose cups dramatically improves communication during an emergency, even with cell phones, radios, or other communication devices.
A NIOSH approved military grade canister provides protection against a range of military agents and NIOSH TRAs (filter data). A HEPA filter provides biological/radiological/particulate protection. This unit also protects against Carbon Monoxide.