Order Entry
United States
ContactUsLinkComponent
CoinLOK™ Coin Bag, Plastic, Clear, 5 mil, CONTROLTEK
CoinLOK™ Coin Bag, Plastic, Clear, 5 mil, CONTROLTEK
Catalog # 76540-066
undefined
CoinLOK™ Coin Bag, Plastic, Clear, 5 mil, CONTROLTEK
Catalog # 76540-066
Supplier Number:  CNK585100

Specifications

  • Color:
    Clear
  • Description:
    Plastic deposit bag with press-to-close glue closure
  • L×W:
    12.5×25"
  • Thickness:
    5 mil.
  • Environmentally Preferable:
  • Cat. No.:
    76540-066

Specifications

About this item

Used every day by major armored couriers and banks, CoinLOK™ provides the convenience of a single-use bag while matching the strength of many reusable bags.

  • Exceptionally strong construction that virtually eliminates breakage
  • Tamper-resistant closure with ultra-strong adhesive for security in transit
  • Reinforced die-cut handle with triple-ply film withstands 100 lbs. stretch test
  • Low slip surface for easy stacking
  • High visibility Fed-compliant barcodes for easy scanning
  • Large serial numbers for easy tracking via CCTV
  • Fed-required labeling and writing areas

CoinLOK™ fits all major coin counters and sorters, improving the efficiency and security of coin processing.

Certifications: Meets VPC (Vault Process Certification) standards for security and performance.