Rhodamine 110 98+%, pure laser grade
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Synonyms: Rhodamine 110 chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid hydrochloride
CAS Number:13558-31-1
:
Formula:C₂₀H₁₅ClN₂O₃
MDL Number:MFCD00042009
Molecular Weight:366.801 g/mol
Melting Pt:>360 °C
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Green to red to red-brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥98.0 %
- UV Lambda max:498 to 504 nm (c=10 mg/l in Methanol)
- Suitability:(for use as a laser dye) Passes test
- Molar absorption coefficient:≥77000 l/(mol.cm) (at Lambda max)
Specification Test Results
Frequently Bought Together