Order Entry
United States
ContactUsLinkComponent
Iron(III) nitrate nonahydrate 99.9995% (trace metals basis)
Iron(III) nitrate nonahydrate 99.9995% (trace metals basis)
Catalog # TS43681-0250
CAS Number:  
undefined
Iron(III) nitrate nonahydrate 99.9995% (trace metals basis)
Catalog # TS43681-0250
Supplier Number:  AC43681-0250
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    25 g
  • Environmentally Preferable:
  • Cat. No.:
    TS43681-0250

Specifications

Specification Test Results

  • Appearance:
    Off-white to violet to purple Crystals
  • Titration Iodometric:
    13.5 to 14.2 % (Fe)
  • Total trace metal impurities:
    ≤5 ppm

Specification Test Results

About this item

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
 :
 :404 g/mol
Boiling Pt:125
UN:1466
 :GHS07
 :GHS03
Storage Temperature:Ambient
CAS Number:7782-61-8
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708
Melting Pt:47
Density:1.68 g/cm³
ADR:5.1 III
Merck Index:13,04057