Membrane Filters with Grid, Sartorius
Supplier: Sartorius
Certificates
About this item
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0.45 µm pore size are tested and released according to ISO 7704.
Specifications
- Catalog No:
- 76500-654
- 76500-652
- 76500-648
- 76500-660
- 76500-658
- 76500-656
- 76500-680
- 76500-674
- 76500-664
- 76500-662
- 76500-684
- 76500-670
- 76500-668
- 76500-666
- Max. Temperature:
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 130 °C
- 180 °C
- 180 °C
- 130 °C
- 130 °C
- 180 °C
- 130 °C
- 130 °C
- 130 °C