Diversey™ Virex TB Disinfectant Cleaner, Lemon Scent, Essendant
Catalog # 76500-272
Supplier: Janitorial Supplies
Specifications
- Capacity:32 oz.
- Environmentally Preferable:
- Cat. No.:76500-272
- Packaging:Bottle
Specifications
About this item
Ready-to-use, quaternary-based hospital-grade disinfectant provides excellent cleaning and deodorizing action in one easy step.
- Quaternary-based, hospital-grade disinfectant cleans and deodorizes
- Meets bloodborne pathogen standards for decontaminating blood and body fluids
- The ready-to-use formula provides excellent, cost-effective cleaning and disinfection in one step
Powerful bactericide, tuberculocide, virucide, and fungicide disinfects surfaces within three minutes of contact. Suitable for use on bathroom fixtures, bathtubs, countertops, sinks, toilet bowls, urinals, and tiles.