Collagen tubes for testing instrumentation and practicing suture technique.
- Evaluate instrument performance
- Student training for suture technique
- Practice end-to-end and end-to-side anastomosis skills
- Practice advanced or basic cardiac, vascular, and general surgery techniques
Vascular Simulation Vessels are 12" long collagen tubes that are ideal for testing new instrumentation, training, and suture practice giving a simulated vascular texture. The vascular simulation vessel provides a true-to-life feel and texture, uniform size, it is less expensive than allographs and ready for testing or training. Tubes are not sterile.