J&W DB-EUPAH Columns, Agilent Technologies
Specifically designed, tested, and optimized for superior performance with all EU-regulated polycyclic aromatic hydrocarbons.
- Specially designed for analysis of EU-regulated PAHs
- Individually tested with application-specific QC test probe mixture
- Great resolution of critical isomers (e.g., benzo(b, j, k)fluroranthenes)
- Superb thermal stability for accurate analysis of high-boiling PAHs such as dibenzopyrenes
- Excellent signal-to-noise ratio
- Optimized column dimensions for proven performance
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W DB-EUPAH columns are designed and tested to provide superior performance for all 16 EU-regulated polycyclic aromatic hydrocarbons (PAHs). The columns deliver excellent resolution of critical PAH isomers, such as benzo(b,j,k)fluoranthene and great thermal stability for accurate analysis of high boiling PAHs, e.g., dibenzopyrenes.
Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency.
USP code - G51
- Catalog No:
- AG122-9632
- AG122-96L2
- AG122-9612
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- USP Code:
- G51
Frequently Bought Together