Air Science USA® Chemical Cabinet Filtration Systems, Air Science USA LLC
Supplier: Air Science
Certificates
About this item
Vent-Box™ ductless filtration systems are designed to protect laboratory personnel from chemical vapors found inside of standalone chemical safety cabinets. The Vent-Box serves as modular ventilation and a filtration system for all chemical safety cabinets.
- Low-profile, completely portable, no ductwork required
- Compact size, easily positioned on shallow safety cabinets
- Modular design permits multiple configurations
- Universal connections allow retrofit to almost all available flammable and safety cabinets on the market
Air Science Vent-Box™ ductless filtration systems are designed to protect laboratory personnel from chemical vapors found inside of stand alone chemical safety cabinets. The Vent-Box serves as modular ventilation and a filtration system for all chemical safety cabinets.
Modular Fume Extractor for Safety Cabinet Ventilation, 115 V 60 Hz with North American Cord set or 230 V 50 Hz with UK Cord set (unless specified). Stand alone filtration unit connects to flamable safety cabinets.
Certifications: UL-C-61010-1, CAN/CSA C22.2 61010-1-12, EN 61010-1:2010. The 230 V units are CE marked
Ordering information: All Units require a filter in order to operate. Filters and accessories are not included with the unit and must be ordered separately by type depending on your chemical handlings.
Delivery: Includes one VB-HOSEKIT-V. Requires MSPP-AP60-001 Main Carbon Filter sold separately