Air Science USA® Airflow Meter for Ductless Fume Hoods, Air Science USA LLC
MSPP-DWYER
- Description:Airflow meter for ductless fume hood
- Environmentally Preferable:
- Cat. No.:MSPP-DWYER
Side wall mounted air flow indicator for ductless fume hoods.
- Flexible mounting
- Easy to read
- Never needs to be calibrated
Velometer, continuous display of face velocity, side wall mounted.