InSolution Caspase Inhibitor VI
Catalog # 101366-092
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- CAS No.:161401-82-7
- Source:
- Species:
- Size:1 mg
- Environmentally Preferable:
- MDL No.:MFCD00016489
- Protein/Peptide Name:Caspase Inhibitor IV
- Molecular Weight:453,46 g/mol
- Molecular Formula:C₂₁H₂₈FN₃O₇
- Cat. No.:101366-092
- Supplier no.:219011-1MG