ContactUsLinkComponent
Cue™ COVID-19 External Control Swabs Pack, Cue Health
Cue™ COVID-19 External Control Swabs Pack, Cue Health
Catalog # 104010-336
Supplier:  CUE HEALTH, INC MS
CAS Number:  
undefined
Cue™ COVID-19 External Control Swabs Pack, Cue Health
Catalog # 104010-336
Supplier:  CUE HEALTH, INC MS
Supplier Number:  C2110
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    COVID-19 External control swabs pack
  • Environmentally Preferable:
  • Cat. No.:
    104010-336
Specifications
About this item
Three (3) Positive Control Swabs and three (3) Negative Control Swabs for use with the Cue COVID-19 Test for Professional Use (sold separately), the Cue Reader (sold separately), and the Cue Health App to check that the Cue COVID-19 Cartridge is working properly.