Order Entry
United States
ContactUsLinkComponent
Rapid Response™ Mononucleosis Antibody Test Cassette, BTNX
Rapid Response™ Mononucleosis Antibody Test Cassette, BTNX
Catalog # 76473-500
Supplier:  Avantor
CAS Number:  
undefined
Rapid Response™ Mononucleosis Antibody Test Cassette, BTNX
Catalog # 76473-500
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mononucleosis test cassette
  • Environmentally Preferable:
  • Cat. No.:
    76473-500

Specifications

About this item

The Rapid Response® Mononucleosis antibody test cassette is a rapid, qualitative test for the detection of infectious mononucleosis heterophile antibodies in whole blood, serum and plasma, to aid in the diagnosis of infectious mononucleosis.

  • Sample: Whole blood/serum/plasma
  • Format: Cassette
  • Specificity: Infectious Mononucleosis Heterophile Antibodies
  • Time to result: 5 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Lateral flow immunoassay
Delivery: Includes test cassettes, buffer, droppers, negative control, positive control, capillary tubes, product insert.
Caution: This test is intended for in vitro diagnostic use only.