Masterflex® L/S® Sanitary Fitting Pump Tubing Assemblies, PharMed® BPT, Cole-Parmer
Supplier: Avantor
Certificates
About this item
These ¹/₂" mini-clamp sanitary fittings are molded onto each end of the assembly for easy implementation into your high-purity system.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Sterilization: Autoclave, EtO, gamma radiation
- Temperature range: –51 to 132 °C (–60 to 270 °F); pumping: –20 to 100 °C (–4 to 212 °F)
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples.
Characteristics: Non-toxic, non-hemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige.
Requires sanitary gaskets for both connections to ensure quality seal.
Frequently Bought Together