Order Entry
United States
ContactUsLinkComponent
Recombinant Albumin, Molecular Biology Grade
Recombinant Albumin, Molecular Biology Grade
Catalog # 76285-184
CAS Number:  
undefined
Recombinant Albumin, Molecular Biology Grade
Catalog # 76285-184
Supplier Number:  B9200S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
  • Size:
    12 mg
  • Environmentally Preferable:
  • Protein/Peptide Name:
    Recombinant albumin
  • Concentration:
    20 mg/ml
  • Grade:
    Molecular biology grade
  • Cat. No.:
    76285-184

Specifications

About this item

Recombinant Albumin, Molecular Biology Grade, is a non-bovine derived albumin that can serve as an alternative to Bovine Serum Albumin (BSA).

Like BSA, it has been shown to prevent adhesion of enzymes to reaction tubes and pipette surfaces. It also stabilizes some proteins during incubation. Choose Recombinant Albumin, when there is a need to avoid BSA. Please inquire about custom formulations with Recombinant Albumin.