Order Entry
United States
ContactUsLinkComponent
Taq Extender PCR Additive, Agilent Technologies
Taq Extender PCR Additive, Agilent Technologies
Catalog # 99902-292
CAS Number:  
undefined
Taq Extender PCR Additive, Agilent Technologies
Catalog # 99902-292
Supplier Number:  600148
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    Taq extender PCR additive
  • Size:
    1000 U
  • Environmentally Preferable:
  • Cat. No.:
    99902-292
  • Supplier no.:
    600148
  • Details:
    Taq extender is used to improve reliability of and yield of conventional PCR. This additive increases efficiency at which Taq performs and also improves PCR amplification of difficult templates and increases reliability and yield of almost any PCR target up to 10 kb in length.

Specifications

About this item

Improves reliability and yield of conventional Taq-based PCR amplifications.

  • Improves length, yield, and reliability of PCR; Enhances efficiency of template extension by Taq DNA polymerase
  • Improves amplification of difficult targets

Taq Extender PCR additive improves the reliability and yield of conventional Taq-based PCR amplifications. Taq Extender additive increases the efficiency of Taq DNA polymerase extension reactions during each cycle of PCR by reducing mismatch pausing, resulting in a greater percentage of completed extension reactions.

Caution: For research use only. Not for use in diagnostic procedures.