First Aid Only™ Surface Wipes, Acme United
Catalog # 76461-934
Supplier: First Aid Only
Specifications
- Description:Surface wipes
- L×W:5.5×5.5"
- Environmentally Preferable:
- Cat. No.:76461-934
- Packaging:Individually packed, 40 pouches/case
Specifications
About this item
Wipes contain 70% isopropyl alcohol and are individually packaged for easy use.
Refill box can be added to a first aid cabinet or be used as a stand-alone refill.