VWR® Adhesive Sealing Strips, PP, 1×8
Supplier: VWR International
Certificates
About this item
1x8 adhesive sealing strips are ideal for use with 1x8 strip-well ELISA plates.
- Ideal for ELISA, incubation and general use
- Seals a single 8-well row of a 96-well plate or an ELISA strip-well plate
- 2.0 mil PP film
- Compatible with both polystyrene and polypropylene plates
- Certified DNase- and RNase-free
- Recommended temperature range from −40 to +120 °C
8 strips are precut on a continuous liner for ease of use.
Frequently Bought Together