Order Entry
United States
ContactUsLinkComponent
Monarch® gDNA Purification Columns, New England Biolabs
Monarch® gDNA Purification Columns, New England Biolabs
Catalog # 76339-786
CAS Number:  
Monarch® gDNA Purification Columns, New England Biolabs
Catalog # 76339-786
Supplier Number:  T3017L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Description:
    Monarch® gDNA purification columns
  • Size:
    100 columns
  • Environmentally Preferable:
  • Cat. No.:
    76339-786

Specifications

About this item

The Monarch® gDNA Purification Columns are a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010) and can be used to purify up to 30 µg of DNA from a wide variety of biological samples.

  • Designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants

DNA can be eluted in as little as 35 μl for and is ready for use in a variety of downstream applications including RT-PCR, RNA Library Prep for NGS, and transfection. Monarch® columns use less plastic than conventional columns, reducing their environmental footprint without compromising performance. They are supplied in quantities of 100, along with 200 Monarch® Collection Tubes II (NEB #T2018).