Water Circulator (Chiller), CF302LA/CF312LA Series, Yamato
Supplier: Yamato Scientific
Certificates
About this item
Powerful and water saving closed cooling system with excellent cooling capacity.
- Environment friendly coolant used for refrigeration
- Compact and space-saving to fit on laboratory tables and limited space
- User-friendly controller
- Easy maintenance of air intake filter
- Rotatable connector which can freely be moved and be set in the desired orientation
- Convenient drainage of waste water
Safety device includes overcurrent ELCB, temperature sensor failure, temperature upper/lower limit alert, temperature upper/lower limit error, refrigerator high pressure cut off switch fan motor protection, circulation pump protection, delay timer for refrigerator protection.
Features calibration offset function which compensates discrepancy between actual liquid temperature and display temperature. Auto resume function allows user to select whether to resume operation or to remain at standby at the time of recovery from power failures.
One year warranty. Manufactured via environmentally conscious methods and remains eco-friendly all throughout its life cycle.
Delivery: Includes 1× condensate drain hose (1 m), 2× hose clamps, 2× hose nozzles (for flexible hose connection).
Specifications
- Catalog No:
- 76550-230
- Temperature Range:
- −20... ~30 °C (no heating function)