Order Entry
United States
ContactUsLinkComponent
alpha-BHC-d6 (a-1,2,3,4,5,6-Hexachlorocyclohexane-d6) Single-Component Organic Standard, SPEX CertiPrep
alpha-BHC-d6 (a-1,2,3,4,5,6-Hexachlorocyclohexane-d6) Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-490
Supplier:  Avantor
CAS Number:  
alpha-BHC-d6 (a-1,2,3,4,5,6-Hexachlorocyclohexane-d6) Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-490
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methanol (P&T)
  • Pack type:
    Ampoule, amber
  • CAS No.:
    86194-41-4
  • Description:
    alpha-BHC-d6 (a-1,2,3,4,5,6-Hexachlorocyclohexane-d6)
  • Size:
    1 ml
  • Environmentally Preferable:
  • Description:
    alpha-BHC-d6 (a-1,2,3,4,5,6-Hexachlorocyclohexane-d6)
  • Cat. No.:
    SXS-490

Specifications

About this item

Single-Component organic standards.

Single-component standards are available as stock items. They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.