Opt-Diss 410 in situ fiber Optic UV Systems for Dissolution Testing, Distek
Supplier: Avantor
Certificates
About this item
No more sampling! The Distek Opt-Diss 410 in-situ fiber optic UV system measures directly in the vessel, eliminating the need for conventional sampling, and with-it consumables like filters, tubing and syringes, saving time, labor, and money.
- Simple and efficient dissolution testing
- Multicomponent analysis
- Rapid and routine data collection
- In-situ UV measurements
- Single software package
- Industry approved
Moving light rather than liquids also allows generating near real-time dissolution data and nearly limitless sample points as frequently as every five seconds.
Certifications: ETL, CE.
Ordering information: Requires Arch probes (not included). Does not include installation or qualification services.
Delivery: Includes spectrograph, CCD detector, dissolution testing software, imbedded PC with Windows X.X OS and one year comprehensive warranty.