Peficitinib ≥98% (by HPLC)
Supplier: Avantor
Certificates
About this item
CAS Number:944118-01-8
:
Formula:C₁₈H₂₂N₄O₂
Molecular Weight:326.40 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together