ContactUsLinkComponent
Nalgene® Validation Bottles, PC, Clean, Certified, Thermo Scientific
Catalog # 76397-860
undefined
Nalgene® Validation Bottles, PC, Clean, Certified, Thermo Scientific
Catalog # 76397-860
Supplier Number:  383500-05
Specifications
  • Sterility:
    Sterile
  • Capacity:
    5 ml
  • Description:
    PC certified clean validation bottles
  • Environmentally Preferable:
  • Cat. No.:
    76397-860
Specifications
About this item
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Polycarbonate construction
  • Sterile
  • Clear finish
  • Certified clean
These bottles can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0.5 EU/ml endotoxin.