ContactUsLinkComponent
HF-Resistant TFE Tracey Spray Chamber for Optima 4300/5300/7300 V, PerkinElmer
HF-Resistant TFE Tracey Spray Chamber for Optima 4300/5300/7300 V, PerkinElmer
Catalog # 10139-486
Supplier:  PerkinElmer
undefined
HF-Resistant TFE Tracey Spray Chamber for Optima 4300/5300/7300 V, PerkinElmer
Catalog # 10139-486
Supplier:  PerkinElmer
Supplier Number:  N0777497
Specifications
  • Description:
    TFE tracey spray chamber for Optima
  • Environmentally Preferable:
  • Cat. No.:
    10139-486
  • Supplier no.:
    N0777497
Specifications
About this item
This TFE Tracey™ spray chamber is used with Optima™ 4300 V, 5300 V, and 7300 V ICP-OES series instruments.
The O-ring-free Tracey spray chamber, made of tetrafluoroethylene (TFE), is designed to be hydrofluoric acid-resistant. It does not have an internal baffle feature. Its precision is considered good and its purity is considered good. They can be teamed with the PolyCon HF-resistant nebulizers to form the ideal chemically inert sample introduction system for ICP-OES.
Made from totally inert PTFE - impervious to organic solvents and strong acids, even HF Established Tracey cyclonic design ensures excellent sensitivity and precision Fast washout enables highest possible sample throughput Incorporates Helix™ nebulizer interface - no dead volume, smooth lock and release mechanism, no O-rings to replace volume: 50 ml.