Male Luer Lock, Needleless Injection Site, for Bioreactors, Chemglass
Catalog # 76333-462
Supplier: Chemglass
Specifications
- Description:Male Luer Lock, needleless injection site, swabable, polycarbonate, silicone
- Environmentally Preferable:
- Cat. No.:76333-462
Specifications
About this item
Male Luer lock with needleless injection site and swabable polycarbonate, silicone with lach samplers for bioreactors.