Vacuum Filtration Adapters with Hose Connection, DWK Life Sciences
Catalog # KT178110-2440
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- [ST] joint:24/40
- Description:Vacuum filtration adapter with hose connection
- Environmentally Preferable:
- Cat. No.:KT178110-2440
- Pluro stopper:#5 or #6
- Supplier no.:178110-2440
Specifications
About this item
Vacuum filtration adapters with hose connection is used for reduced pressure filtration using glass vessel with standard taper joint.
- Designed for pluro stopper
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Pluro stopper and funnel sold separately.