Flame Resistant Insulated Bib Overalls, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics.
- Available in regular and tall formats
- Max Comfort™ 7.5 oz., 88% cotton/12% nylon outer shell
- 11.5 oz. quilted lining, 100% FR treated cotton and non-woven modacrylic batting
- Max Comfort™ material, sewn with FR aramid thread
- Double needle stitching on main seams
The insulated overall features a comfortable 88% cotton/12% nylon material with a modacrylic quilted lining for warmth and protection. Max Comfort™ material meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace.
Certifications: Max Comfort™ fabric NFPA 2112 UL certified. All trims are NFPA 2112 UL certified. ASTM F1506 electric arc protection standard; NFPA 70E hazard/risk Cat HRC3; ATPV ASTM 1959 35 cal/cm², HRC 3; bib overall is UL and NFPA 2112 certified.
Specifications
- Catalog No:
- 76342-152
- 76342-368
- 76342-366
- 76341-572
- 76342-408
- 76342-406
- 76342-450
- 76342-324
- 76342-440
- 76342-182
- 76342-430
- Garment application:
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- Fabric:
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- 88% cotton/12% nylon outer shell
- Weight:
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
Specifications
Frequently Bought Together