The original Jamshidi™ needle offers features that have made this bone marrow biopsy/aspiration needle a best seller for decades. The needles are available in a wide variety of sizes to meet user preference. All needles are packaged with an adapter to accommodate luer lock syringes.
- Razor-sharp bevel tip allows easy coring of the bone
- Single-use design helps assure sterility for every procedure and helps prevent cross contamination
Lightweight handle with built-in comfort knob enables the practitioner to apply sufficient pressure without discomfort. Tapered distal tip facilitates the recovery of bone marrow samples with unaltered architecture.