Dichloroacetic acid (DCA) 3% in anhydrous dichloromethane deblocking reagent, for biosynthesis, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Synonyms: DCA Deblock, DCA
CAS Number:79-43-6
:
Formula:C₂H₂Cl₂O₂
MDL Number:MFCD00004223
Boiling Pt:194
Molecular Weight:128.94 g/mol
Melting Pt:-4
ADR:8 II
Merck Index:13,03075
UN:2922
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Description (clear liquid free of suspended matter):Pass
- Dichloroacetic Acid:2.95 - 3.05%
- Color (at time of packaging):<10 APHA
- Water:Max 100 ppm
- Dichloromethane (component purity):Min 99.9%
- Dicloroacetic Acid (component purity):Min 98.0%
Specification Test Results
Frequently Bought Together