ContactUsLinkComponent
PHCbi MPR Series Upright Pharmaceutical Refrigerators, PHC Corporation
PHCbi MPR Series Upright Pharmaceutical Refrigerators, PHC Corporation
Supplier:  PHC Corporation
   Sustainable Options Available
Certificates
undefined
PHCbi MPR Series Upright Pharmaceutical Refrigerators, PHC Corporation
Supplier:  PHC Corporation
   Sustainable Options Available
Specifications
  • Catalog No:
  • 76355-506
  • 76355-508
  • Model:
  • MPR-722-PA
  • MPR-722R-PA
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Cat. No.:
  • 76355-506
  • 76355-508
  • Display:
  • LED (displayed by 1, 0.1)
  • LED (displayed by 1, 0.1)
  • Alarm:
  • Visual, buzzer and remote alarm
  • Visual, buzzer and remote alarm
  • Access Ports:
  • 3 (Left, Right, Top) with Ø 30 mm (1.1")
  • 3 (Left, Right, Top) with Ø 30 mm (1.1")
  • Shelves/drawers:
  • 4 adjustable shelves
  • 5 solid pull-out drawers
  • Material:
  • Galvanised steel
  • Galvanised steel
  • Electrical:
  • 115 V, 60 Hz
  • 115 V, 60 Hz
  • Weight:
  • 174 kg (334 lbs.)
  • 193 kg (425 lbs.)