SYPRO® Ruby Protein Blot Stain, Lonza
Catalog # 12001-746
Supplier: Lonza
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:SYPRO® Ruby Protein Blot stain
- Size:200 ml (6.8 oz.)
- Environmentally Preferable:
- Cat. No.:12001-746
- Supplier no.:50565
Specifications
About this item
SYPRO™ Ruby is 60-times more sensitive than reversible protein blot stains like Ponceau S, and 30-times more sensitive than Amido Black or Coomassie® brilliant blue stains.
- Visualize proteins prior to transfer - does not interfere with protein activity
- Versatile - Excellent for staining native proteins
- Safe - No acids or organic solvents necessary
- Sensitive - Detects as little as 4 to 8 ng protein per band