To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:Engelbreth-Holm-Swarm (EHS) mouse tumor
- Species:
- Description:Matrigel® Matrix for organoid culture
- Size:10 ml
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:ISO 14001 - Bedford (reg. no. 14.4942)
- Storage Temperature:–20 °C
- Cat. No.:76332-052
Specifications
About this item
Corning Matrigel Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.
- Quality tested for ability to form and maintain stable 3D domes/droplets on multi-well plates
- Elastic Modulus (matrix stiffness) measured using a rheometer
- Protein concentrations determined by Lowry method
- Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
- Extensive PCR testing is performed to screen for a number of pathogens
- Tested and found negative for bacteria, fungi, and mycoplasma
- Endotoxin units measured by Limulus Amoebocyte Lysate assay
Verified to support both mouse and human organoid growth and differentiation from healthy and/or diseased tissue, each lot has a specific elastic modulus value indicating the stiffness of the gel formed and is qualified to form stable 3D domes commonly used in organoid culture protocols. Available in 10 ml LDEV-free, Phenol Red-free format.
Corning Matrigel Matrix for Organoid culture is extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.