Statfree® High Density Conductive Foam, Desco
Supplier: Desco
Certificates
About this item
Antistatic, low tribocharging dissipative foam is corrosion resistant and ideal for long term storage.
- Uniform protection
- Lead-free RoHS compliant
Antistatic, low tribocharging foam minimizes charge generation. It will conduct charges if grounded. Uniform protection will not affect the solderability of device leads.
Certifications: Surface resistance: 1×10³ to <1×10⁶ ohms per ANSI/ESD STM11.11. S. Volume resistance: 1×10³ to <1×10⁶ ohms per ANSI/ESD STM11.12.
Frequently Bought Together