Flowmeters, Bubble, Chemglass
Supplier: Chemglass
Certificates
About this item
Flowmeter used for the control of carrier gasses.
- Projection at top breaks bubbles
Hose connection has a outer diameter of 9 mm at the middle ring.
Delivery: Supplied complete with rubber bulb.
Frequently Bought Together