ContactUsLinkComponent
Rubbermaid® Commercial Clean Room Maintenance Mop Heads
Rubbermaid® Commercial Clean Room Maintenance Mop Heads
Catalog # 66103-130
undefined
Rubbermaid® Commercial Clean Room Maintenance Mop Heads
Catalog # 66103-130
Supplier Number:  RCPT300
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    66103-130
  • Supplier no.:
    RCP T300
  • Headband size:
    5"
  • Mop head size:
    Medium
Specifications
About this item
Sturdy, looped-end tailbanded floor mop for industrial applications, including pharmaceutical labs, paint booths, and more.
  • Made of launderable, pre-shrunk rayon/polyester fabric that will not fray or lint
  • Headband color: Green
  • Mop head color: White