ContactUsLinkComponent
Nuclisens® easyMAG® Disposable, Biomerieux
Nuclisens® easyMAG® Disposable, Biomerieux
Catalog # 89130-520
Supplier:  Avantor
Nuclisens® easyMAG® Disposable, Biomerieux
Catalog # 89130-520
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    NUCLISENS® easyMAG® Disposable
  • Size:
    1 Set (48 pcs)
  • Environmentally Preferable:
  • Cat. No.:
    89130-520
  • Supplier no.:
    280135
  • Sufficient for:
    8 extractions
Specifications
About this item
The NUCLISENS® easyMAG® accessory products are reagents and disposables for automated extraction (purification and concentration) of total nucleic acids (RNA/DNA) from homogeneous human biological fluid samples, to be used with the bioMerieux instruments.
  • 16 x 3 sample vessels and 3 aspirator disposables
  • Plastic disposables with a row of sample wells.
With NUCLISENS easyMAG, perform quality total nucleic acid extraction as integral part of your molecular diagnostic operations. This system demonstrates great flexibility as it can be used with a variety of sample types and sample volumes.
Also used on EMAG® system