Molecular sieve A5 (0.5 nm, 5 Å), pellets, Fluka™
BJ334316-1KG
:
- Pack type:Plastic bottle
- Size:1 kg
- Environmentally Preferable:
- Particle Size:1.6 mm
- Form:pellets
- Granulation:complying
- Pore Size (Ång Ström):5
: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
:
:69912-79-4
:1318-02-1