Proteomelab™ SDS-MW Analysis Kit, SCIEX
Catalog # MSPP-390953
Supplier: AB SCIEX LLC MS
CAS Number:
Specifications
- Description:Proteomelab SDS-MW Analysis kit
- Size:1 kit
- Environmentally Preferable:
- Cat. No.:MSPP-390953
Specifications
About this item
The SDS-Gel MW analysis kit is designed for the separation and sizing of protein–SDS complexes using a replaceable gel matrix. The gel is formulated to provide an effective protein sieving range of approximately 10 to 225 kDa.
Used to effectively quantify the amount of protein and to determine the purity of a protein product.
Delivery: The kit contains separation capillary 50 µm I.D. bare fused silica (2), SDS-MW gel buffer - proprietary formulation, pH 8, 0.2% SDS (140 ml, 4 pack), SDS-MW sample buffer - 100 mM Tris-HCl, pH 9.0/1% SDS (50 ml), SDS-MW size standard 10 kDa to 225 kDa, 16 mg/ml (100 µl), internal standard, 10 kDa protein, 5 mg/ml, (0.4 ml), acidic wash solution, 0.1 M HCl (100 ml), basic wash solution, 0.1 M NaOH (100 ml), SDS-Gel MW analysis guide (1).
Caution: The SDS-MW analysis kit is for laboratory use only.