Infusion Kits, Analytical Sales and Services
Supplier: ANALYTICAL SALES & SERVICE MS
Certificates
About this item
Ready to use infusion kits.
Delivery: Includes one syringe, adapter, fittings, tubing, PEEK™ tee connection, and one standard replacement needle.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.