Transparent Bath Tanks For Use With Heating Immersion Circulator To +100 °C, JULABO USA, Inc.
Supplier: JULABO
Certificates
About this item
Transparent Plexiglass bath tanks.
- Clear Plexiglass tanks
- Rim with integrated handles
- Suitable for use with CORIO C/CD heating immersion circulators
Transparent bath tanks for sample incubation in conjunction with heating immersion circulators. Tank volumes available in 9, 19 and 27 l sizes.
Frequently Bought Together